Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,3-Diphenyl-2-cyclopropen-1-one, 99% (dry wt.), may cont up to 5% water

Click to view available options
Quantity:
1g
5g
25g
886-38-4
C15H10O
206.244
MFCD00001311
HCIBTBXNLVOFER-UHFFFAOYSA-N
diphenylcyclopropenone,diphencyprone,2,3-diphenylcyclopropenone,2,3-diphenylcycloprop-2-enone,cyclopropenone, diphenyl,dpcp,2-cyclopropen-1-one, 2,3-diphenyl,1,2-diphenylcyclopropen-3-one,cyclopropenone, 2,3-diphenyl,unii-i7g14nw5ec
65057
CHEBI:53074
2,3-diphenylcycloprop-2-en-1-one
C1=CC=C(C=C1)C2=C(C2=O)C3=CC=CC=C3
886-38-4
C15H10O
206.244
MFCD00001311
HCIBTBXNLVOFER-UHFFFAOYSA-N
diphenylcyclopropenone,diphencyprone,2,3-diphenylcyclopropenone,2,3-diphenylcycloprop-2-enone,cyclopropenone, diphenyl,dpcp,2-cyclopropen-1-one, 2,3-diphenyl,1,2-diphenylcyclopropen-3-one,cyclopropenone, 2,3-diphenyl,unii-i7g14nw5ec
65057
CHEBI:53074
2,3-diphenylcycloprop-2-en-1-one
C1=CC=C(C=C1)C2=C(C2=O)C3=CC=CC=C3
Melting Point 119°C to 121°C
Quantity 5g
Beilstein 608049
Merck Index 14,3308
Formula Weight 206.24
Percent Purity 99%
Chemical Name or Material 2,3-Diphenyl-2-cyclopropen-1-one
Assay (dry wt.), may cont up to 5% water

GHS H Statement
H317
May cause an allergic skin reaction.

GHS P Statement
P261-P280-P363-P321-P333+P313-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash contaminated clothing before reuse.
Specific treatment (see label).
If skin irritation or rash occurs: Get medical advice/attention.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning