Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Diphenylmaleic anhydride, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
4808-48-4
C16H10O3
250.26
OUJCFCNZIUTYBH-UHFFFAOYSA-N
2,3-diphenylmaleic anhydride,diphenylmaleic anhydride,3,4-diphenyl-2,5-furandione,diphenylfuran-2,5-dione,2,3-diphenylmaleicanhydride,2,5-furandione, 3,4-diphenyl,acmc-1asck,2, 3,4-diphenyl,diphenyl-maleic anhydride,2,5-furandione,3,4-diphenyl
78530
3,4-diphenylfuran-2,5-dione
C1=CC=C(C=C1)C2=C(C(=O)OC2=O)C3=CC=CC=C3
4808-48-4
C16H10O3
250.26
OUJCFCNZIUTYBH-UHFFFAOYSA-N
2,3-diphenylmaleic anhydride,diphenylmaleic anhydride,3,4-diphenyl-2,5-furandione,diphenylfuran-2,5-dione,2,3-diphenylmaleicanhydride,2,5-furandione, 3,4-diphenyl,acmc-1asck,2, 3,4-diphenyl,diphenyl-maleic anhydride,2,5-furandione,3,4-diphenyl
78530
3,4-diphenylfuran-2,5-dione
C1=CC=C(C=C1)C2=C(C(=O)OC2=O)C3=CC=CC=C3
Color Yellow
Quantity 5g
Infrared Spectrum Authentic
Melting Point 157.1°C to 159.4°C
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Formula Weight 250.26
Physical Form Crystalline Powder or Powder
Percent Purity 98%
Chemical Name or Material 2, 3-Diphenylmaleic anhydride
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Co

GHS Signal Word: Warning

EINECSNumber : 225-370-2