Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Diphenylpropionic Acid 98.0+%, TCI America™
SDP

Catalog No. D179025G
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 3333-15-1
Molecular Formula C15H14O2
Molecular Weight (g/mol) 226.28
MDL Number MFCD00044405
InChI Key PCRICPYPVZKEBZ-UHFFFAOYNA-N
PubChem CID 18744
IUPAC Name 2,3-diphenylpropanoic acid
SMILES OC(=O)C(CC1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 89°C
Color White-Yellow
Boiling Point 340°C
Quantity 25 g
Formula Weight 226.28
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,3-Diphenylpropionic Acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.