missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-(3-Fluorophenyl)ethylamine, ≥97%

missing translation for 'orderingAttributeHoverText'
Quantity:
5g
25g
100g
404-70-6
C8H10FN
139.173
MFCD00075376
AUCVZEYHEFAWHO-UHFFFAOYSA-N
3-fluorophenethylamine,2-3-fluorophenyl ethylamine,2-3-fluoro-phenyl-ethylamine,2-3-fluorophenyl ethanamine,2-3-fluorophenyl ethan-1-amine,benzeneethanamine, 3-fluoro,m-fluorophenethylamine,3-flourophenethylamine,m-fluorophenylethylamine,3-fluoro phenethylamine
533928
2-(3-fluorophenyl)ethanamine
C1=CC(=CC(=C1)F)CCN
404-70-6
C8H10FN
139.173
MFCD00075376
AUCVZEYHEFAWHO-UHFFFAOYSA-N
3-fluorophenethylamine,2-3-fluorophenyl ethylamine,2-3-fluoro-phenyl-ethylamine,2-3-fluorophenyl ethanamine,2-3-fluorophenyl ethan-1-amine,benzeneethanamine, 3-fluoro,m-fluorophenethylamine,3-flourophenethylamine,m-fluorophenylethylamine,3-fluoro phenethylamine
533928
2-(3-fluorophenyl)ethanamine
C1=CC(=CC(=C1)F)CCN
Quantity 100g
Boiling Point 87°C (15mmHg)
Density 1.066g/mL
Refractive Index 1.5090
UN Number UN2735
Beilstein 3045789
Formula Weight 139.17
Physical Form Liquid
Percent Purity ≥97%
Chemical Name or Material 2-(3-Fluorophenyl)ethylamine

GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packaging Group: II; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.