Alfa Aesar™ 2-(3-Fluorophenylamino)benzoic acid, 98%

Manufacturer: Alfa Aesar™ 4385204

Quantity 2g

 View more versions of this product

Catalog No. AA4385204

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 54-59-1
    Molecular Formula C13H10FNO2
    Formula Weight 231.22
    MDL Number MFCD01675228
    Synonym N-(3-Fluorophenyl)anthranilic acid
    Chemical Name or Material 2- (3-Fluorophenylamino)benzoic acid

    Specifications

    Formula Weight 231.22
    MDL Number MFCD01675228
    Synonym N-(3-Fluorophenyl)anthranilic acid
    Physical Form Powder
    Molecular Formula C13H10FNO2
    Melting Point 158° to 163°C
    CAS 54-59-1
    Color Pale yellow to gray
    Quantity 2g
    RTECS Number CB3060000
    Chemical Name or Material 2- (3-Fluorophenylamino)benzoic acid
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit