Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Heptanedione 98.0+%, TCI America™

Click to view available options
Quantity:
5mL
25mL
96-04-8
C7H12O2
128.171
MFCD00036550
FJPGAMCQJNLTJC-UHFFFAOYSA-N
2,3-heptanedione,acetyl valeryl,valerylacetyl,acetylvaleryl,acetyl pentanoyl,unii-dk55dde86p,dk55dde86p,benzil-related compound, 47,heptanedione,fema no. 2543
60983
heptane-2,3-dione
CCCCC(=O)C(=O)C
96-04-8
C7H12O2
128.171
MFCD00036550
FJPGAMCQJNLTJC-UHFFFAOYSA-N
2,3-heptanedione,acetyl valeryl,valerylacetyl,acetylvaleryl,acetyl pentanoyl,unii-dk55dde86p,dk55dde86p,benzil-related compound, 47,heptanedione,fema no. 2543
60983
heptane-2,3-dione
CCCCC(=O)C(=O)C
Boiling Point 64°C
Color Yellow
Quantity 25mL
UN Number 1224
Formula Weight 128.17
Physical Form Clear Liquid at 20°C
Percent Purity ≥98.0% (GC)
Chemical Name or Material 2,3-Heptanedione

EINECSNumber : (2)-0563

TSCA : Yes

Recommended Storage : Refrigerator