Alfa Aesar™ 2-(3-Methoxyphenyl)cyclohexanone, tech. 90%

Manufacturer: Alfa Aesar™ B2261903

Quantity 1g

 View more versions of this product

Catalog No. AAB2261903

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 15547-89-4
    Molecular Formula C13H16O2
    Formula Weight 204.27
    MDL Number MFCD00001630
    Chemical Name or Material 2-(3-Methoxyphenyl)cyclohexanone
    Grade Technical Grade

    Specifications

    Formula Weight 204.27
    Quantity 1g
    Molecular Formula C13H16O2
    MDL Number MFCD00001630
    Boiling Point 140° to 142°C (0.2mmHg)
    CAS 15547-89-4
    Flash Point >110°C (230°F)
    Chemical Name or Material 2-(3-Methoxyphenyl)cyclohexanone
    Assay 90%
    Grade Technical Grade

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit