Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-(3-Methoxyphenyl)cyclohexanone, tech. 90%

Click to view available options
Quantity:
1g
5g
25g
15547-89-4
C13H16O2
204.269
MFCD00001630
KACQSVYTBQDRGP-UHFFFAOYSA-N
2-3-methoxyphenyl cyclohexanone,2-3-methoxyphenyl cyclohexan-1-one,cyclohexanone, 2-3-methoxyphenyl,acmc-20aoq7,2-3-methoxyphenyl-cyclohexanone,2-3-methoxy-phenyl-cyclohexanone,cyclohexanone,2-3-methoxyphenyl,2-3-methoxyphenyl-cyclohexan-1-one,2-3-methoxyphenyl cyclohexanone, technical grade
85868
2-(3-methoxyphenyl)cyclohexan-1-one
COC1=CC=CC(=C1)C2CCCCC2=O
15547-89-4
C13H16O2
204.269
MFCD00001630
KACQSVYTBQDRGP-UHFFFAOYSA-N
2-3-methoxyphenyl cyclohexanone,2-3-methoxyphenyl cyclohexan-1-one,cyclohexanone, 2-3-methoxyphenyl,acmc-20aoq7,2-3-methoxyphenyl-cyclohexanone,2-3-methoxy-phenyl-cyclohexanone,cyclohexanone,2-3-methoxyphenyl,2-3-methoxyphenyl-cyclohexan-1-one,2-3-methoxyphenyl cyclohexanone, technical grade
85868
2-(3-methoxyphenyl)cyclohexan-1-one
COC1=CC=CC(=C1)C2CCCCC2=O
Quantity 5g
Boiling Point 140°C to 142°C (0.2mmHg)
Flash Point >110°C (230°F)
Formula Weight 204.27
Percent Purity 90%
Grade Technical
Chemical Name or Material 2-(3-Methoxyphenyl)cyclohexanone