Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-(3-Methylphenyl)acetamidoxime, 97%

Click to view available options
Quantity:
250mg
1g
885952-60-3
C9H12N2O
164.208
PUBSVUGAWXVFMA-UHFFFAOYSA-N
benzeneethanimidamide,n-hydroxy-3-methyl,n-hydroxy-2-m-tolyl-acetamidine,n'-hydroxy-2-3-methylphenyl ethanimidamide,2-3-tolyl-acetamidoxime,n-hydroxy-2-m-tolyl acetimidamide
24688408
N'-hydroxy-2-(3-methylphenyl)ethanimidamide
CC1=CC(=CC=C1)CC(=NO)N
885952-60-3
C9H12N2O
164.208
PUBSVUGAWXVFMA-UHFFFAOYSA-N
benzeneethanimidamide,n-hydroxy-3-methyl,n-hydroxy-2-m-tolyl-acetamidine,n'-hydroxy-2-3-methylphenyl ethanimidamide,2-3-tolyl-acetamidoxime,n-hydroxy-2-m-tolyl acetimidamide
24688408
N'-hydroxy-2-(3-methylphenyl)ethanimidamide
CC1=CC(=CC=C1)CC(=NO)N
Quantity 1g
Color Brown to White
Formula Weight 164.2
Physical Form Crystalline Solid
Percent Purity 97%
Chemical Name or Material 2-(3-Methylphenyl)acetamidoxime