Alfa Aesar™ (+)-2,3-O-Isopropylidene-L-threitol, 98%

Manufacturer: Alfa Aesar™ B2309003

Quantity 1g

 View more versions of this product

Catalog No. AAB2309003

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 50622-09-8
    Molecular Formula C7H14O4
    Formula Weight 162.19
    MDL Number MFCD00063761
    Beilstein 1280797
    Synonym (4S,5S)-2,2-Dimethyl-1,3-dioxolane-4,5-dimethanol
    Chemical Name or Material (+)-2,3-O-Isopropylidene-L-threitol

    Specifications

    Formula Weight 162.19
    Molecular Formula C7H14O4
    Synonym (4S,5S)-2,2-Dimethyl-1,3-dioxolane-4,5-dimethanol
    Boiling Point 92° to 94°C (0.1mmHg)
    Flash Point >110°C (230°F)
    Quantity 1g
    MDL Number MFCD00063761
    CAS 50622-09-8
    Beilstein 1280797
    Melting Point 45° to 49°C
    Optical Rotation +3° (c=5 in Ethanol)
    Chemical Name or Material (+)-2,3-O-Isopropylidene-L-threitol
    Assay 98%

    Description

    Applications
    (+)-2,3-O-Isopropylidene-L-threitol is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

    Solubility
    Soluble in water.

    Notes
    Hygroscopic. Store away from oxidizing agents, water/ moisture.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit