Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(3-Phenylpropyl)pyridine, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
2110-18-1
C14H15N
197.28
MFCD00191237
JJJPNTQYUJPWGQ-UHFFFAOYSA-N
2-3-phenylpropyl pyridine,pyridine, 2-3-phenylpropyl,unii-9f0bfm744f,alpha-3-phenylpropyl pyridine,2-phenylpropylpyridine,cortex pyridine,phenylpropylpyridine,3-phenylpropyl pyridine,acmc-1cs6o,dsstox_cid_22356
459494
2-(3-phenylpropyl)pyridine
C1=CC=C(C=C1)CCCC2=CC=CC=N2
2110-18-1
C14H15N
197.28
MFCD00191237
JJJPNTQYUJPWGQ-UHFFFAOYSA-N
2-3-phenylpropyl pyridine,pyridine, 2-3-phenylpropyl,unii-9f0bfm744f,alpha-3-phenylpropyl pyridine,2-phenylpropylpyridine,cortex pyridine,phenylpropylpyridine,3-phenylpropyl pyridine,acmc-1cs6o,dsstox_cid_22356
459494
2-(3-phenylpropyl)pyridine
C1=CC=C(C=C1)CCCC2=CC=CC=N2
Specific Gravity 1.015
Quantity 1g
Density 1.0150g/mL
Color Yellow
Packaging Glass bottle
Infrared Spectrum Authentic
Refractive Index 1.5590 to 1.5610 (20°C, 589nm)
Flash Point 100°C
Boiling Point 115.0°C (2.0 mmHg)
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Formula Weight 197.28
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 2-(3-Phenylpropyl)pyridine
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 218-300-7