Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-[3-(Trifluoromethyl)phenyl]acetamidoxime, 97%, Alfa Aesar™

Click to view available options
Quantity:
250mg
1g
925393-51-7
925393-51-7
Quantity 250mg
Color White
Formula Weight 218.18
Physical Form Solid
Percent Purity 97%
Chemical Name or Material 2-[3-(Trifluoromethyl)phenyl]acetamidoxime