Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2'-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)furan 97%, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
374790-93-9
C10H15BO3
194.037
MFCD02094008
GWEWQKZABZXLJH-UHFFFAOYSA-N
2-furanboronic acid pinacol ester,furan-2-boronic acid pinacol ester,2-2-furanyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane,2-furan-2-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane,furan-2-boronic acid pinacol cyclic ester,2-2-furyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane,2'-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl furan,1,3,2-dioxaborolane, 2-2-furanyl-4,4,5,5-tetramethyl,zlchem 693,pubchem18440
11356013
2-(furan-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
B1(OC(C(O1)(C)C)(C)C)C2=CC=CO2
374790-93-9
C10H15BO3
194.037
MFCD02094008
GWEWQKZABZXLJH-UHFFFAOYSA-N
2-furanboronic acid pinacol ester,furan-2-boronic acid pinacol ester,2-2-furanyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane,2-furan-2-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane,furan-2-boronic acid pinacol cyclic ester,2-2-furyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane,2'-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl furan,1,3,2-dioxaborolane, 2-2-furanyl-4,4,5,5-tetramethyl,zlchem 693,pubchem18440
11356013
2-(furan-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
B1(OC(C(O1)(C)C)(C)C)C2=CC=CO2
Quantity 5g
Packaging Glass bottle
Color Colorless to Yellow
Infrared Spectrum Authentic
Refractive Index 1.4730 to 1.4750 (20°C, 589nm)
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Formula Weight 194.04
Physical Form Liquid
Percent Purity ≥96.0%
Chemical Name or Material 2'-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)furan
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.