Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4,4-Trimethyl-1-pentene, 99%, ACROS Organics™

Click to view available options
Quantity:
100mL
500mL
Packaging:
Glass bottle
107-39-1
C8H16
112.21
MFCD00008855
FXNDIJDIPNCZQJ-UHFFFAOYSA-N
2,4,4-trimethyl-1-pentene,diisobutylene,pentene, 2,4,4-trimethyl,2,4,4-trimethylpentene,1-pentene, 2,4,4-trimethyl,2,2,4-trimethyl-4-pentene,unii-n69l73advf,1-methyl-1-neopentylethylene,ccris 9103,2,4,4-trimethylpentene-1
7868
2,4,4-trimethylpent-1-ene
CC(=C)CC(C)(C)C
107-39-1
C8H16
112.21
MFCD00008855
FXNDIJDIPNCZQJ-UHFFFAOYSA-N
2,4,4-trimethyl-1-pentene,diisobutylene,pentene, 2,4,4-trimethyl,2,4,4-trimethylpentene,1-pentene, 2,4,4-trimethyl,2,2,4-trimethyl-4-pentene,unii-n69l73advf,1-methyl-1-neopentylethylene,ccris 9103,2,4,4-trimethylpentene-1
7868
2,4,4-trimethylpent-1-ene
CC(=C)CC(C)(C)C
Refractive Index 1.4070 to 1.409
Density 0.7000g/mL
Melting Point -101°C
Boiling Point 101°C to 102°C
Specific Gravity 0.7
Flash Point -6°C
Color Colorless
Packaging Glass bottle
Quantity 500mL
Infrared Spectrum Authentic
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula (CH3)3CCH2C(CH3)=CH2
Solubility Information Solubility in water: immiscible with water
Formula Weight 112.21
Physical Form Liquid
Percent Purity ≥98.5%
Chemical Name or Material 2, 4, 4-Trimethyl-1-pentene
Show More Show Less

GHS H Statement:
Toxic to aquatic life with long lasting effects.
Highly flammable liquid and vapor.

GHS P Statement:
Avoid release to the environment.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger