Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4,5,6-Tetrachloropyrimidine, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
1780-40-1
C4Cl4N2
217.87
MFCD00006062
GVBHCMNXRKOJRH-UHFFFAOYSA-N
perchloropyrimidine,pyrimidine, tetrachloro,tetrachloropyrimidine,pyrimidine, 2,4,5,6-tetrachloro,2,4,5,6-tetrachloro-1,3-pyrimidine,tetrachlorpyrimidin,tetrachloro-pyrimidin,acmc-1bsuq,2,5,6-tetrachloropyrimidine,5-23-05-00347 beilstein handbook reference
15690
2,4,5,6-tetrachloropyrimidine
C1(=C(N=C(N=C1Cl)Cl)Cl)Cl
1780-40-1
C4Cl4N2
217.87
MFCD00006062
GVBHCMNXRKOJRH-UHFFFAOYSA-N
perchloropyrimidine,pyrimidine, tetrachloro,tetrachloropyrimidine,pyrimidine, 2,4,5,6-tetrachloro,2,4,5,6-tetrachloro-1,3-pyrimidine,tetrachlorpyrimidin,tetrachloro-pyrimidin,acmc-1bsuq,2,5,6-tetrachloropyrimidine,5-23-05-00347 beilstein handbook reference
15690
2,4,5,6-tetrachloropyrimidine
C1(=C(N=C(N=C1Cl)Cl)Cl)Cl
Melting Point 67°C to 68.5°C
Packaging Glass Bottle
Boiling Point 108°C to 112°C (12.0mmHg)
Infrared Spectrum Authentic
Color White
Quantity 25g
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Formula Weight 217.87
Physical Form Crystalline Powder
Percent Purity 97%
Chemical Name or Material 2,4,5,6-Tetrachloropyrimidine
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

GHS Signal Word: Warning

EINECSNumber : 217-228-3

RTECSNumber : UV9745000

TSCA : TSCA