Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,4,5-Triamino-6-hydroxypyrimidine sulfate, 97%

Manufacturer:  Alfa Aesar™ B2198922

Quantity 100g

 View more versions of this product

Catalog No. AAB2198922


  Check Availability
Add to cart

Specifications

Description

Applications
2,4,5-Triamino-6-hydroxypyrimidine sulfate used as an intermediate for synthetic organic chemistry.

Solubility
Soluble in DMSO.

Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents.
Description & Specifications

Chemical Identifiers

35011-47-3
239.21
4621048
2,4,5-Triamino-6-hydroxypyrimidine sulfate
C4H7N5O·H2SO4
MFCD00012785
6-Hydroxy-2,4,5-triaminopyrimidine sulfate; 2,5,6-Triamino-4(3H)-pyrimidinone sulfate

Specifications

>300°C
35011-47-3
239.21
4621048
2,4,5-Triamino-6-hydroxypyrimidine sulfate
100g
C4H7N5O·H2SO4
MFCD00012785
6-Hydroxy-2,4,5-triaminopyrimidine sulfate; 2,5,6-Triamino-4(3H)-pyrimidinone sulfate
97%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : UW7175000

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit