Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4,5-Tribromoimidazole, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
100g
Packaging:
Glass bottle
2034-22-2
C3HBr3N2
304.76
MFCD00005184
JCGGPCDDFXIVQB-UHFFFAOYSA-N
2,4,5-tribromoimidazole,1h-imidazole, 2,4,5-tribromo,imidazole, 2,4,5-tribromo,2,4,5-tribromoimidazo,tribromoimidazole,pubchem7150,2,5-tribromoimidazole,acmc-1cmbx,timtec-bb sbb001252,imidazole,4,5-tribromo
16253
2,4,5-tribromo-1H-imidazole
C1(=C(N=C(N1)Br)Br)Br
2034-22-2
C3HBr3N2
304.76
MFCD00005184
JCGGPCDDFXIVQB-UHFFFAOYSA-N
2,4,5-tribromoimidazole,1h-imidazole, 2,4,5-tribromo,imidazole, 2,4,5-tribromo,2,4,5-tribromoimidazo,tribromoimidazole,pubchem7150,2,5-tribromoimidazole,acmc-1cmbx,timtec-bb sbb001252,imidazole,4,5-tribromo
16253
2,4,5-tribromo-1H-imidazole
C1(=C(N=C(N1)Br)Br)Br
Packaging Glass bottle
Color Beige-Yellow
Quantity 5g
Infrared Spectrum Authentic
Melting Point 208.0°C to 212.0°C
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Beilstein 23, 49
Formula Weight 304.76
Physical Form Powder
Percent Purity 97%
Chemical Name or Material 2, 4, 5-Tribromoimidazole
Show More Show Less

GHS H Statement:
Toxic if swallowed.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 217-997-5