Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4,5-Trichlorobenzenesulfonyl chloride 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
15945-07-0
C6H2Cl4O2S
279.94
MFCD00007428
WNVVRCKTQSCPAC-UHFFFAOYSA-N
2,4,5-trichlorobenzene-1-sulfonyl chloride,benzenesulfonyl chloride, 2,4,5-trichloro,unii-7gd5283xix,2,4,5-trichlorobenzenesulphonyl chloride,2,4,5-trichlorobenzenesulfonylchloride,pubchem5174,acmc-1c50t,2,4,5trichlorophenylsulfonylchloride,2,5-trichlorobenzenesulfonyl chloride,chloro 2,4,5-trichlorophenyl sulfone
27575
2,4,5-trichlorobenzenesulfonyl chloride
C1=C(C(=CC(=C1Cl)Cl)Cl)S(=O)(=O)Cl
15945-07-0
C6H2Cl4O2S
279.94
MFCD00007428
WNVVRCKTQSCPAC-UHFFFAOYSA-N
2,4,5-trichlorobenzene-1-sulfonyl chloride,benzenesulfonyl chloride, 2,4,5-trichloro,unii-7gd5283xix,2,4,5-trichlorobenzenesulphonyl chloride,2,4,5-trichlorobenzenesulfonylchloride,pubchem5174,acmc-1c50t,2,4,5trichlorophenylsulfonylchloride,2,5-trichlorobenzenesulfonyl chloride,chloro 2,4,5-trichlorophenyl sulfone
27575
2,4,5-trichlorobenzenesulfonyl chloride
C1=C(C(=CC(=C1Cl)Cl)Cl)S(=O)(=O)Cl
Quantity 25g
Packaging Glass bottle
Infrared Spectrum Authentic
Color Brown to Yellow
Melting Point 66°C to 70°C
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula Cl3C6H2SO2Cl
Formula Weight 279.94
Physical Form Liquid
Percent Purity ≥96.0% (GC)
Chemical Name or Material 2, 4, 5-Trichlorobenzenesulfonyl chloride
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Reacts violently with water.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger