Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,4,5-Trichlorophenol, 94%

Manufacturer:  Alfa Aesar™ 4373722

Quantity 100g

 View more versions of this product

Catalog No. AA4373722

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

95-95-4
197.45
UN2020
14,9643
C6H3Cl3O
MFCD00002170
607569
2,4,5-Trichlorophenol

Specifications

100g
1.678g/mL
C6H3Cl3O
MFCD00002170
607569
Phenol-like
67°C
94%
95-95-4
Powder
197.45
UN2020
14,9643
253°C
2,4,5-Trichlorophenol

Safety and Handling

GHS H Statement
H302-H315-H319
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P273-P305+P351+P338-P302+P352-P501a
Avoid release to the environment.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF ON SKIN: Wash with plenty of soap and water.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CHLOROPHENOLS, SOLID

RTECSNumber : SN1400000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit