Alfa Aesar™ 2,4,5-Trichlorophenol, 94%

Manufacturer: Alfa Aesar™ 43737-14

Quantity 25g

 View more versions of this product

Catalog No. AA4373714

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 95-95-4
    Molecular Formula C6H3Cl3O
    Formula Weight 197.45
    MDL Number MFCD00002170
    UN Number UN2020
    Beilstein 607569
    Merck Index 14,9643
    Chemical Name or Material 2,4,5-Trichlorophenol

    Specifications

    Formula Weight 197.45
    Health Hazard 3 GHS P Statement
    P273-P305+P351+P338-P302+P352-P501a
    Avoid release to the environment.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF ON SKIN: Wash with plenty of soap and water.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 1 Warning
    Odor Phenol-like
    Molecular Formula C6H3Cl3O
    Beilstein 607569
    Merck Index 14,9643
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CHLOROPHENOLS, SOLID
    Quantity 25g
    TSCA TSCA
    MDL Number MFCD00002170
    CAS 95-95-4
    UN Number UN2020
    Health Hazard 2 GHS H Statement
    H302-H315-H319
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    Boiling Point 253°C
    Melting Point 67°C
    Density 1.678g/mL
    Physical Form Powder
    RTECS Number SN1400000
    Chemical Name or Material 2,4,5-Trichlorophenol
    Assay 94%

    GHS H Statement
    H302-H315-H319
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P273-P305+P351+P338-P302+P352-P501a
    Avoid release to the environment.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF ON SKIN: Wash with plenty of soap and water.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit