Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4,5-Trichloropyrimidine, 98%, Acros Organics™

Click to view available options
Quantity:
1g
5g
25g
5750-76-5
C4HCl3N2
183.42
GIKMWFAAEIACRF-UHFFFAOYSA-N
2,5,6-trichloropyrimidine,2,4,5-trichloro-pyrimidine,trichloropyrimidine,pyrimidine, 2,4,5-trichloro,2,4,5-trichlorpyrimidin,pubchem5302,zlchem 1349,pyrimidine, trichloro,acmc-209lys,2,5-trichloropyrimidine
237259
2,4,5-trichloropyrimidine
C1=C(C(=NC(=N1)Cl)Cl)Cl
5750-76-5
C4HCl3N2
183.42
GIKMWFAAEIACRF-UHFFFAOYSA-N
2,5,6-trichloropyrimidine,2,4,5-trichloro-pyrimidine,trichloropyrimidine,pyrimidine, 2,4,5-trichloro,2,4,5-trichlorpyrimidin,pubchem5302,zlchem 1349,pyrimidine, trichloro,acmc-209lys,2,5-trichloropyrimidine
237259
2,4,5-trichloropyrimidine
C1=C(C(=NC(=N1)Cl)Cl)Cl
Packaging Glass Bottle
Flash Point >110°C
Infrared Spectrum Authentic
Quantity 1g
Assay Percent Range 97.5% min. (GC)
Formula Weight 183.42
Percent Purity 98%
Chemical Name or Material 2,4,5-Trichloropyrimidine

GHS H Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Call a POISON CENTER or doctor/physician if you feel unwell.

GHS Signal Word: Warning