Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4,5-Trichlorothiophenol, 90%, Tech., ACROS Organics™

Click to view available options
Quantity:
10g
Packaging:
Glass bottle
3773-14-6
C6H3Cl3S
213.5
JARIALSGFXECCH-UHFFFAOYSA-N
2,4,5-trichlorothiophenol,renacit ii,benzenethiol, 2,4,5-trichloro,2,4,5-trichlorobenzene-1-thiol,acmc-209iv9,2,4,5-trichloro-thiophenol,2,4,5-trichlorophenyl mercaptan,2,4,5-tris chloranyl benzenethiol,n-memet-n-trimetsilylmet benzylamine
19597
2,4,5-trichlorobenzenethiol
C1=C(C(=CC(=C1Cl)Cl)Cl)S
3773-14-6
C6H3Cl3S
213.5
JARIALSGFXECCH-UHFFFAOYSA-N
2,4,5-trichlorothiophenol,renacit ii,benzenethiol, 2,4,5-trichloro,2,4,5-trichlorobenzene-1-thiol,acmc-209iv9,2,4,5-trichloro-thiophenol,2,4,5-trichlorophenyl mercaptan,2,4,5-tris chloranyl benzenethiol,n-memet-n-trimetsilylmet benzylamine
19597
2,4,5-trichlorobenzenethiol
C1=C(C(=CC(=C1Cl)Cl)Cl)S
Quantity 10g
Infrared Spectrum Authentic
Packaging Glass bottle
Color White to Beige
CAS Min % 88.0
CAS Max % 100.0
Assay Percent Range 90%
Linear Formula Cl3C6H2SH
Formula Weight 213.51
Physical Form Crystalline Powder
Percent Purity ≥88%
Grade Technical
Chemical Name or Material 2, 4, 5-Trichlorothiophenol, tech., 90%
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning