Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4,5-Trifluorobenzaldehyde, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
165047-24-5
C7H3F3O
160.09
MFCD00061196
CYIFJRXFYSUBFW-UHFFFAOYSA-N
2,4,5-trifluoro benzaldehyde,benzaldehyde, 2,4,5-trifluoro,benzaldehyde, 2,4,5-trifluoro-9ci,pubchem1468,pubchem4258,intermediates-zcf02238,acmc-209ds3,2,4,5-trifluorbenzaldehyde,2,4,5-trifluorobenzoaldehyde,ksc494i3d
519278
2,4,5-trifluorobenzaldehyde
C1=C(C(=CC(=C1F)F)F)C=O
165047-24-5
C7H3F3O
160.09
MFCD00061196
CYIFJRXFYSUBFW-UHFFFAOYSA-N
2,4,5-trifluoro benzaldehyde,benzaldehyde, 2,4,5-trifluoro,benzaldehyde, 2,4,5-trifluoro-9ci,pubchem1468,pubchem4258,intermediates-zcf02238,acmc-209ds3,2,4,5-trifluorbenzaldehyde,2,4,5-trifluorobenzoaldehyde,ksc494i3d
519278
2,4,5-trifluorobenzaldehyde
C1=C(C(=CC(=C1F)F)F)C=O
Specific Gravity 1.408
Color Colorless to Yellow
Infrared Spectrum Authentic
Density 1.4080g/mL
Quantity 1g
Flash Point 58°C
Boiling Point 168.0°C
Packaging Glass bottle
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Linear Formula F3C6H3CHO
Formula Weight 160.09
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material 2, 4, 5-Trifluorobenzaldehyde
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning