Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4,5-Trifluorobenzenesulfonyl chloride, 97%, Acros Organics

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
220227-21-4
C6H2ClF3O2S
230.59
STCZWXXRRTWRSK-UHFFFAOYSA-N
2,4,5-trifluorobenzene-1-sulfonyl chloride,2,4,5-trifluorobenzenesulphonyl chloride,2,4,5-trifluorophenyl sulfonyl chloride,2,4,5-trifluoro-benzenesulfonyl chloride,benzenesulfonyl chloride, 2,4,5-trifluoro,pubchem5179,acmc-209fqs,ksc201o4t,chloro 2,4,5-trifluorophenyl sulfone,2,4,5-trifluorophenylsulfonyl chloride
2776979
2,4,5-trifluorobenzenesulfonyl chloride
C1=C(C(=CC(=C1F)S(=O)(=O)Cl)F)F
220227-21-4
C6H2ClF3O2S
230.59
STCZWXXRRTWRSK-UHFFFAOYSA-N
2,4,5-trifluorobenzene-1-sulfonyl chloride,2,4,5-trifluorobenzenesulphonyl chloride,2,4,5-trifluorophenyl sulfonyl chloride,2,4,5-trifluoro-benzenesulfonyl chloride,benzenesulfonyl chloride, 2,4,5-trifluoro,pubchem5179,acmc-209fqs,ksc201o4t,chloro 2,4,5-trifluorophenyl sulfone,2,4,5-trifluorophenylsulfonyl chloride
2776979
2,4,5-trifluorobenzenesulfonyl chloride
C1=C(C(=CC(=C1F)S(=O)(=O)Cl)F)F
Specific Gravity 1.657
Packaging Glass bottle
Quantity 1g
Flash Point >100°C
Density 1.6570g/mL
Assay Percent Range 96% min. (GC)
Linear Formula F3C6H2SO2Cl
Formula Weight 230.59
Percent Purity 97%
Chemical Name or Material 2, 4, 5-Trifluorobenzenesulfonyl chloride

GHS H Statement
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately

GHS Signal Word: Danger