Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,4,5-Trifluorobenzyl bromide, 98%

Click to view available options
Quantity:
1g
5g
157911-56-3
C7H4BrF3
225.008
MFCD00061209
GAUXEYCSWSMMFZ-UHFFFAOYSA-N
2,4,5-trifluorobenzyl bromide,1-bromomethyl-2,4,5-trifluorobenzene,2,4,5-trifluorobenzylbromide,2,4,5-trifluoro benzyl bromide,benzene, 1-bromomethyl-2,4,5-trifluoro,alpha-bromo-2,4,5-trifluorotoluene,2, 4, 5-trifluorobenzyl bromide,pubchem3026,1-bromomethyl-2,4,5-trifluoro-benzene,acmc-1c0kl
2777057
1-(bromomethyl)-2,4,5-trifluorobenzene
C1=C(C(=CC(=C1F)F)F)CBr
157911-56-3
C7H4BrF3
225.008
MFCD00061209
GAUXEYCSWSMMFZ-UHFFFAOYSA-N
2,4,5-trifluorobenzyl bromide,1-bromomethyl-2,4,5-trifluorobenzene,2,4,5-trifluorobenzylbromide,2,4,5-trifluoro benzyl bromide,benzene, 1-bromomethyl-2,4,5-trifluoro,alpha-bromo-2,4,5-trifluorotoluene,2, 4, 5-trifluorobenzyl bromide,pubchem3026,1-bromomethyl-2,4,5-trifluoro-benzene,acmc-1c0kl
2777057
1-(bromomethyl)-2,4,5-trifluorobenzene
C1=C(C(=CC(=C1F)F)F)CBr
Quantity 5g
Refractive Index 1.508
UN Number UN3265
Beilstein 8312091
Formula Weight 225.01
Percent Purity 98%
Chemical Name or Material 2,4,5-Trifluorobenzyl bromide

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures