Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,4,5-Trifluorobenzyl chloride, 97%

Click to view available options
Quantity:
1g
5g
243139-71-1
C7H4ClF3
180.554
MFCD00236319
JMXPOOVDUVHJRO-UHFFFAOYSA-N
2,4,5-trifluorobenzyl chloride,1-chloromethyl-2,4,5-trifluorobenzene,2,4,5-trifluorobenzylchloride,2,4,5-trifluorobenzyl choride,benzene, 1-chloromethyl-2,4,5-trifluoro,alpha-chloro-2,4,5-trifluorotoluene,1-chloromethyl-2,4,5-trifluoro-benzene,pubchem4024,acmc-1cjlx
2777063
1-(chloromethyl)-2,4,5-trifluorobenzene
C1=C(C(=CC(=C1F)F)F)CCl
243139-71-1
C7H4ClF3
180.554
MFCD00236319
JMXPOOVDUVHJRO-UHFFFAOYSA-N
2,4,5-trifluorobenzyl chloride,1-chloromethyl-2,4,5-trifluorobenzene,2,4,5-trifluorobenzylchloride,2,4,5-trifluorobenzyl choride,benzene, 1-chloromethyl-2,4,5-trifluoro,alpha-chloro-2,4,5-trifluorotoluene,1-chloromethyl-2,4,5-trifluoro-benzene,pubchem4024,acmc-1cjlx
2777063
1-(chloromethyl)-2,4,5-trifluorobenzene
C1=C(C(=CC(=C1F)F)F)CCl
Refractive Index 1.505
Density 1.689
Flash Point 65°C (150°F)
Quantity 1g
UN Number UN3265
Formula Weight 180.56
Percent Purity 97%
Chemical Name or Material 2,4,5-Trifluorobenzyl chloride

GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.

P210-P235-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c

H227-H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures