Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,4,5-Trimethoxybenzaldehyde, 98%

Click to view available options
Quantity:
25g
100g
4460-86-0
C10H12O4
196.202
MFCD00003312
IAJBQAYHSQIQRE-UHFFFAOYSA-N
asaraldehyde,asaronaldehyde,asarylaldehyde,azarylaldehyde,benzaldehyde, 2,4,5-trimethoxy,gazarin,3,4,6-trimethoxybenzaldehyde,unii-ndu8j2q00d,2,4,5-trimethoxy-benzaldehyde,ccris 1296
20525
2,4,5-trimethoxybenzaldehyde
COC1=CC(=C(C=C1C=O)OC)OC
4460-86-0
C10H12O4
196.202
MFCD00003312
IAJBQAYHSQIQRE-UHFFFAOYSA-N
asaraldehyde,asaronaldehyde,asarylaldehyde,azarylaldehyde,benzaldehyde, 2,4,5-trimethoxy,gazarin,3,4,6-trimethoxybenzaldehyde,unii-ndu8j2q00d,2,4,5-trimethoxy-benzaldehyde,ccris 1296
20525
2,4,5-trimethoxybenzaldehyde
COC1=CC(=C(C=C1C=O)OC)OC
Quantity 100g
Melting Point 111°C to 115°C
Boiling Point 140°C (4mmHg)(sublimation)
Beilstein 1951403
Formula Weight 196.2
Percent Purity 98%
Chemical Name or Material 2,4,5-Trimethoxybenzaldehyde

RTECSNumber : CU8460000

TSCA : TSCA