Alfa Aesar™ 2,4,5-Trimethyl-3-oxazoline, cis + trans, 99%

Manufacturer: Alfa Aesar™ B2222814

Quantity 25g

 View more versions of this product

Catalog No. AAB2222814

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 22694-96-8
    Molecular Formula C6H11NO
    Formula Weight 113.16
    MDL Number MFCD00053161
    UN Number UN1993
    Chemical Name or Material 2,4,5-Trimethyl-3-oxazoline, cis + trans

    Specifications

    Formula Weight 113.16
    MDL Number MFCD00053161
    UN Number UN1993
    Health Hazard 1 Warning
    Health Hazard 3 GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    RTECS Number RQ5665000
    Odor Characteristic
    Boiling Point 45° to 47°C (12mmHg)
    Quantity 25g
    Molecular Formula C6H11NO
    CAS 22694-96-8
    Health Hazard 2 GHS H Statement
    H226-H315-H319-H335
    Flammable liquid and vapour.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
    Chemical Name or Material 2,4,5-Trimethyl-3-oxazoline, cis + trans
    Assay 99%

    Description

    Applications
    2,4,5-Trimethyl-3-oxazoline, cis + trans possesses the characteristic meat aroma hences used for flavoring application. Used as flavour enhancer in fish

    Solubility
    Soluble in alcohol, fixed oils. Insoluble in water.

    Notes
    Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air.

    GHS H Statement
    H226-H315-H319-H335
    Flammable liquid and vapour.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit