Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ 2,4,5-Trimethyl-3-oxazoline, cis + trans, 99%
Click to view available options
Quantity:
5g
25g
100g
Description
Applications
2,4,5-Trimethyl-3-oxazoline, cis + trans possesses the characteristic meat aroma hences used for flavoring application. Used as flavour enhancer in fish
Solubility
Soluble in alcohol, fixed oils. Insoluble in water.
Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air.
2,4,5-Trimethyl-3-oxazoline, cis + trans possesses the characteristic meat aroma hences used for flavoring application. Used as flavour enhancer in fish
Solubility
Soluble in alcohol, fixed oils. Insoluble in water.
Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air.
Specifications
Sensitivity | Air Sensitive |
Odor | Characteristic |
Quantity | 5g |
Boiling Point | 45°C to 47°C (12mmHg) |
UN Number | UN1993 |
Solubility Information | Soluble in alcohol,fixed oils. Insoluble in water. |
Formula Weight | 113.16 |
Percent Purity | 99% |
Chemical Name or Material | 2,4,5-Trimethyl-3-oxazoline, cis + trans |
Safety and Handling
GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H226-H315-H319-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
RTECSNumber : RQ5665000
TSCA : No
Recommended Storage : Ambient temperatures; Store under Argon