Alfa Aesar™ 2',4',5'-Trimethylacetophenone, 97%

Manufacturer: Alfa Aesar™ A1061418

Quantity 50g

 View more versions of this product

Catalog No. AAA1061418

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 2040-07-5
    Molecular Formula C11H14O
    Formula Weight 162.23
    MDL Number MFCD00017244
    Beilstein 2042909
    Chemical Name or Material 2',4',5'-Trimethylacetophenone

    Specifications

    Formula Weight 162.23
    CAS 2040-07-5
    Molecular Formula C11H14O
    MDL Number MFCD00017244
    Melting Point 10° to 11°C
    Quantity 50g
    Beilstein 2042909
    Boiling Point 246° to 248°C
    Chemical Name or Material 2',4',5'-Trimethylacetophenone
    Assay 97%

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit