Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,4,5-Triphenylimidazole, 97%

Click to view available options
Quantity:
10g
50g
250g
484-47-9
C21H16N2
296.373
MFCD00005187
RNIPJYFZGXJSDD-UHFFFAOYSA-N
2,4,5-triphenylimidazole,lophine,lophin,1h-imidazole, 2,4,5-triphenyl,2,4,5-triphenyl-1,3-imidazole,unii-q6k46g80zd,imidazole, 2,4,5-triphenyl,chembl34173,2,5-triphenylimidazole,imidazole,4,5-triphenyl
10232
2,4,5-triphenyl-1H-imidazole
C1=CC=C(C=C1)C2=C(N=C(N2)C3=CC=CC=C3)C4=CC=CC=C4
484-47-9
C21H16N2
296.373
MFCD00005187
RNIPJYFZGXJSDD-UHFFFAOYSA-N
2,4,5-triphenylimidazole,lophine,lophin,1h-imidazole, 2,4,5-triphenyl,2,4,5-triphenyl-1,3-imidazole,unii-q6k46g80zd,imidazole, 2,4,5-triphenyl,chembl34173,2,5-triphenylimidazole,imidazole,4,5-triphenyl
10232
2,4,5-triphenyl-1H-imidazole
C1=CC=C(C=C1)C2=C(N=C(N2)C3=CC=CC=C3)C4=CC=CC=C4
Applications
2,4,5-Triphenylimidazole is used in the synthesis of fluorescent diarylethenes and nanowires of 2,4,5-triphenylimidazole, which finds application as single wire active optical waveguides and optically driven ultraviolet lasers.

Solubility
Soluble in methanol. Insoluble in water.

Notes
Incompatible with strong oxidizing agents, strong acids and bases. Store in a cool place.
Quantity 250g
Melting Point 274°C to 278°C
Beilstein 236652
Solubility Information Soluble in methanol. Insoluble in water.
Formula Weight 296.38
Percent Purity 97%
Chemical Name or Material 2,4,5-Triphenylimidazole

EINECSNumber : 207-606-6

RTECSNumber : NI8710000

TSCA : Yes

Recommended Storage : Ambient temperatures