Learn More
2,4,6-Collidine, 99%, Thermo Scientific Chemicals
Description
2,4,6-Collidine is used as a tissue fixative for electron microscopy. It is useful in dehydrohalogenation reactions and acts as a solvent for the cleavage of hindered esters by anhydrous lithium iodide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
2,4,6-Collidine is used as a tissue fixative for electron microscopy. It is useful in dehydrohalogenation reactions and acts as a solvent for the cleavage of hindered esters by anhydrous lithium iodide.
Solubility
Slightly miscible with water. Miscible with ether, methanol, acetone, chloroform, benzene, toluene and dilute acids.
Notes
Incompatible with oxidizing agents and strong acids.
Specifications
Melting Point | -43°C |
Density | 0.916 |
pH | 9 to 10 |
Boiling Point | 171°C to 172°C |
Flash Point | 54°C (129°F) |
Odor | Unpleasant |
Refractive Index | 1.498 |
Quantity | 25 mL |
UN Number | UN1992 |
Beilstein | 107283 |
Show More |
Safety and Handling
Hazard Category | H226-H302+H332-H311-H315-H319-H335 |
Hazard Statement | GHS H Statement H311-H226-H302-H315-H319-H335 Toxic in contact with skin. Flammable liquid and vapor. Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c |
DOTInformation | Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, TOXIC, N.O.S. |
EINECSNumber | 203-613-3 |
RTECSNumber | UU0970000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.