Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4,6-Tri-tert-butylnitrosobenzene, 99%, ACROS Organics™

Click to view available options
Quantity:
250mg
1g
Packaging:
Glass bottle
24973-59-9
C18H29NO
275.436
MFCD00008817
OSICDPWAPKXXHT-UHFFFAOYSA-N
2,4,6-tri-tert-butylnitrosobenzene,1,3,5-tri-tert-butyl-2-nitrosobenzene,2,4,6-tri-t-butyl-nitrosobenzene,ttbnb,1,3,5-tri tert-butyl-2-nitrosobenzene,benzene, 1,3,5-tris 1,1-dimethylethyl-2-nitroso,2-nitroso-1,3,5-tri-tert-butylbenzene,benzene,1,3,5-tris 1,1-dimethylethyl-2-nitroso,acmc-1cr76,2,4,6-tri-t-butylnitrosobenzene
90676
1,3,5-tritert-butyl-2-nitrosobenzene
CC(C)(C)C1=CC(=C(C(=C1)C(C)(C)C)N=O)C(C)(C)C
24973-59-9
C18H29NO
275.436
MFCD00008817
OSICDPWAPKXXHT-UHFFFAOYSA-N
2,4,6-tri-tert-butylnitrosobenzene,1,3,5-tri-tert-butyl-2-nitrosobenzene,2,4,6-tri-t-butyl-nitrosobenzene,ttbnb,1,3,5-tri tert-butyl-2-nitrosobenzene,benzene, 1,3,5-tris 1,1-dimethylethyl-2-nitroso,2-nitroso-1,3,5-tri-tert-butylbenzene,benzene,1,3,5-tris 1,1-dimethylethyl-2-nitroso,acmc-1cr76,2,4,6-tri-t-butylnitrosobenzene
90676
1,3,5-tritert-butyl-2-nitrosobenzene
CC(C)(C)C1=CC(=C(C(=C1)C(C)(C)C)N=O)C(C)(C)C
Melting Point 170°C to 173°C
Infrared Spectrum Authentic
Packaging Glass bottle
Color Green
Quantity 250mg
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula {(CH3)3C}3C6H2NO
Formula Weight 275.42
Physical Form Powder
Percent Purity ≥98.5%
Chemical Name or Material 2, 4, 6-Tri-tert-butylnitrosobenzene, 99%
Show More Show Less

EINECSNumber : 246-553-3

TSCA : TSCA