Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4,6-Tri-tert-butylphenol, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
50g
500g
Packaging:
Glass Bottle
Plastic bottle
732-26-3
C18H30O
262.42
MFCD00008821
PFEFOYRSMXVNEL-UHFFFAOYSA-N
2,4,6-tri-tert-butylphenol,voidox,alkofen b,2,4,6-tris tert-butyl phenol,2,4,6-tri-t-butylphenol,phenol, 2,4,6-tris 1,1-dimethylethyl,phenol, 2,4,6-tri-tert-butyl,2,4,6-tris 1,1-dimethylethyl phenol,unii-99j2vic675,tris 1,1-dimethylethyl phenol
12902
2,4,6-tritert-butylphenol
CC(C)(C)C1=CC(=C(C(=C1)C(C)(C)C)O)C(C)(C)C
732-26-3
C18H30O
262.42
MFCD00008821
PFEFOYRSMXVNEL-UHFFFAOYSA-N
2,4,6-tri-tert-butylphenol,voidox,alkofen b,2,4,6-tris tert-butyl phenol,2,4,6-tri-t-butylphenol,phenol, 2,4,6-tris 1,1-dimethylethyl,phenol, 2,4,6-tri-tert-butyl,2,4,6-tris 1,1-dimethylethyl phenol,unii-99j2vic675,tris 1,1-dimethylethyl phenol
12902
2,4,6-tritert-butylphenol
CC(C)(C)C1=CC(=C(C(=C1)C(C)(C)C)O)C(C)(C)C
Melting Point 129°C to 132°C
Boiling Point 277°C
Packaging Plastic bottle
Quantity 500g
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula [(CH3)3C]3C6H2OH
UN Number 3077
Beilstein 06, IV, 3539
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in most common organic solvents
Formula Weight 262.42
Chemical Name or Material 2, 4, 6-Tri-tert-butylphenol
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Very toxic to aquatic life with long lasting effects.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning