Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4,6-Triaminopyrimidine, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass Bottle
Plastic bottle
1004-38-2
C4H7N5
125.13
MFCD00006100
JTTIOYHBNXDJOD-UHFFFAOYSA-N
2,4,6-triaminopyrimidine,2,4,6-pyrimidinetriamine,pyrimidine-2,4,6-triyltriamine,pyrimidine, 2,4,6-triamino,triamino pyrimidine,3ay,aaat,pubchem7216,2,6-pyrimidinetriamine,2,6-triaminopyrimidine
13863
CHEBI:39857
pyrimidine-2,4,6-triamine
C1=C(N=C(N=C1N)N)N
1004-38-2
C4H7N5
125.13
MFCD00006100
JTTIOYHBNXDJOD-UHFFFAOYSA-N
2,4,6-triaminopyrimidine,2,4,6-pyrimidinetriamine,pyrimidine-2,4,6-triyltriamine,pyrimidine, 2,4,6-triamino,triamino pyrimidine,3ay,aaat,pubchem7216,2,6-pyrimidinetriamine,2,6-triaminopyrimidine
13863
CHEBI:39857
pyrimidine-2,4,6-triamine
C1=C(N=C(N=C1N)N)N
Quantity 5g
Packaging Glass Bottle
Infrared Spectrum Authentic
Color White to Beige
Melting Point 246°C to 252°C
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Beilstein 24,47
Solubility Information Solubility in water: 36.5g/kg in water (20°C). Other solubilities: 22.7g/kg in methanol (20°C),4.5g/kg in ethanol (20°C)
Formula Weight 125.13
Physical Form Powder
Percent Purity 97%
Chemical Name or Material 2, 4, 6-Triaminopyrimidine, 97%
Show More Show Less

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 213-720-7

TSCA : TSCA