Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4,6-Tribromoaniline 98.0+%, TCI America™

Click to view available options
Quantity:
25g
500g
147-82-0
C6H4Br3N
329.817
MFCD00007634
GVPODVKBTHCGFU-UHFFFAOYSA-N
benzenamine, 2,4,6-tribromo,s-tribromoaniline,2,4,6-tribromophenylamine,2,4,6-tribromobenzenamine,sym-tribromoaniline,aniline tribromide,tribromoaniline,usaf do-43,aniline, 2,4,6-tribromo,2,4,6-tribomoaniline
8986
2,4,6-tribromoaniline
C1=C(C=C(C(=C1Br)N)Br)Br
147-82-0
C6H4Br3N
329.817
MFCD00007634
GVPODVKBTHCGFU-UHFFFAOYSA-N
benzenamine, 2,4,6-tribromo,s-tribromoaniline,2,4,6-tribromophenylamine,2,4,6-tribromobenzenamine,sym-tribromoaniline,aniline tribromide,tribromoaniline,usaf do-43,aniline, 2,4,6-tribromo,2,4,6-tribomoaniline
8986
2,4,6-tribromoaniline
C1=C(C=C(C(=C1Br)N)Br)Br
Quantity 500g
Color White
Melting Point 120°C
Formula Weight 329.82
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (GC)
Chemical Name or Material 2,4,6-Tribromoaniline

EINECSNumber : (3)-0292

RTECSNumber : BZ0175000

TSCA : Yes