Alfa Aesar™ 2,4,6-Tribromoanisole, 99%

Manufacturer: Alfa Aesar™ H5568414

Quantity 25g

 View more versions of this product

Catalog No. AAH5568414

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 607-99-8
    Formula Weight 344.83
    MDL Number MFCD00192510
    Beilstein 2210361
    Chemical Name or Material 2,4,6-Tribromoanisole

    Specifications

    Formula Weight 344.83
    Melting Point 84° to 88°C
    Boiling Point 297° to 299°C
    Beilstein 2210361
    Color White to pale brown
    Physical Form Solid
    MDL Number MFCD00192510
    CAS 607-99-8
    TSCA TSCA
    Quantity 25g
    Chemical Name or Material 2,4,6-Tribromoanisole
    Assay 99%

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit