Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4,6-Trichlorobenzoyl chloride, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
4136-95-2
C7H2Cl4O
243.9
OZGSEIVTQLXWRO-UHFFFAOYSA-N
2,4,6-trichlorobenzoylchloride,benzoyl chloride, 2,4,6-trichloro,mi,acmc-209jk3,ksc494s2p,2,4,6-cl3c6h2cocl,2,4,6-ttrichlorobenzoyl chloride,2,4,6-trichloro benzoyl chloride,2,4,6-trichlorobenzoic acid chloride,2,4,6-tris chloranyl benzoyl chloride
2733703
2,4,6-trichlorobenzoyl chloride
C1=C(C=C(C(=C1Cl)C(=O)Cl)Cl)Cl
4136-95-2
C7H2Cl4O
243.9
OZGSEIVTQLXWRO-UHFFFAOYSA-N
2,4,6-trichlorobenzoylchloride,benzoyl chloride, 2,4,6-trichloro,mi,acmc-209jk3,ksc494s2p,2,4,6-cl3c6h2cocl,2,4,6-ttrichlorobenzoyl chloride,2,4,6-trichloro benzoyl chloride,2,4,6-trichlorobenzoic acid chloride,2,4,6-tris chloranyl benzoyl chloride
2733703
2,4,6-trichlorobenzoyl chloride
C1=C(C=C(C(=C1Cl)C(=O)Cl)Cl)Cl
Flash Point >110°C
Boiling Point 107.0°C to 108.0°C (6.0 mmHg)
Infrared Spectrum Authentic
Quantity 5g
Packaging Glass bottle
Specific Gravity 1.56
Density 1.5600g/mL
Refractive Index 1.5750 to 1.5770
Color Yellow
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Merck Index 15,9798
Formula Weight 243.9
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 2, 4, 6-Trichlorobenzoyl chloride
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger