Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4,6-Trichlorobenzoyl chloride, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
4136-95-2
C7H2Cl4O
243.9
OZGSEIVTQLXWRO-UHFFFAOYSA-N
2,4,6-trichlorobenzoylchloride,benzoyl chloride, 2,4,6-trichloro,mi,acmc-209jk3,ksc494s2p,2,4,6-cl3c6h2cocl,2,4,6-ttrichlorobenzoyl chloride,2,4,6-trichloro benzoyl chloride,2,4,6-trichlorobenzoic acid chloride,2,4,6-tris chloranyl benzoyl chloride
2733703
2,4,6-trichlorobenzoyl chloride
C1=C(C=C(C(=C1Cl)C(=O)Cl)Cl)Cl
4136-95-2
C7H2Cl4O
243.9
OZGSEIVTQLXWRO-UHFFFAOYSA-N
2,4,6-trichlorobenzoylchloride,benzoyl chloride, 2,4,6-trichloro,mi,acmc-209jk3,ksc494s2p,2,4,6-cl3c6h2cocl,2,4,6-ttrichlorobenzoyl chloride,2,4,6-trichloro benzoyl chloride,2,4,6-trichlorobenzoic acid chloride,2,4,6-tris chloranyl benzoyl chloride
2733703
2,4,6-trichlorobenzoyl chloride
C1=C(C=C(C(=C1Cl)C(=O)Cl)Cl)Cl
Quantity 25g
Color Yellow
Infrared Spectrum Authentic
Flash Point >110°C
Boiling Point 107.0°C to 108.0°C (6.0 mmHg)
Density 1.5600g/mL
Packaging Glass bottle
Refractive Index 1.5750 to 1.5770
Specific Gravity 1.56
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Merck Index 15,9798
Formula Weight 243.9
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 2, 4, 6-Trichlorobenzoyl chloride
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger