Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4,6-Trichloronitrobenzene 96.0+%, TCI America™

Click to view available options
Quantity:
5g
25g
18708-70-8
C6H2Cl3NO2
226.437
MFCD00014690
AEBJDOTVYMITIA-UHFFFAOYSA-N
2,4,6-trichloronitrobenzene,benzene, 1,3,5-trichloro-2-nitro,2,4,6-trichloro-1-nitrobenzene,unii-t00kp6ct7k,t00kp6ct7k,benzene,1,3,5-trichloro-2-nitro,acmc-209epa,2,6-trichloronitrobenzene,dsstox_cid_6204,dsstox_rid_78056
29228
1,3,5-trichloro-2-nitrobenzene
C1=C(C=C(C(=C1Cl)[N+](=O)[O-])Cl)Cl
18708-70-8
C6H2Cl3NO2
226.437
MFCD00014690
AEBJDOTVYMITIA-UHFFFAOYSA-N
2,4,6-trichloronitrobenzene,benzene, 1,3,5-trichloro-2-nitro,2,4,6-trichloro-1-nitrobenzene,unii-t00kp6ct7k,t00kp6ct7k,benzene,1,3,5-trichloro-2-nitro,acmc-209epa,2,6-trichloronitrobenzene,dsstox_cid_6204,dsstox_rid_78056
29228
1,3,5-trichloro-2-nitrobenzene
C1=C(C=C(C(=C1Cl)[N+](=O)[O-])Cl)Cl
Quantity 25g
Melting Point 72°C
Color White-Yellow
Formula Weight 226.44
Physical Form Crystal-Powder at 20°C
Percent Purity ≥96.0% (GC)
Chemical Name or Material 2,4,6-Trichloronitrobenzene

EINECSNumber : (3)-0459

TSCA : Yes