Alfa Aesar™ 2,4,6-Trichlorophenol, 98%

Manufacturer: Alfa Aesar™ A116810I

Quantity 5000g

 View more versions of this product

Catalog No. AAA116810I

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 88-06-2
    Molecular Formula C6H3Cl3O
    Formula Weight 197.45
    MDL Number MFCD00002172
    UN Number UN2020
    Beilstein 776729
    Merck Index 14,9644
    Chemical Name or Material 2,4,6-Trichlorophenol

    Specifications

    Formula Weight 197.45
    Assay Percent Range 98%
    Health Hazard 3 GHS P Statement
    P280-P281-P305+P351+P338-P362-P405-P501a
    Wear protective gloves/protective clothing/eye protection/face protection.
    Use personal protective equipment as required.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Take off contaminated clothing and wash before reuse.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    TSCA TSCA
    Health Hazard 2 GHS H Statement
    H351-H302-H315-H319
    Suspected of causing cancer.
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    Molecular Formula C6H3Cl3O
    Boiling Point 245° to 246°C
    Melting Point 63° to 68°C
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CHLOROPHENOLS, SOLID
    MDL Number MFCD00002172
    Beilstein 776729
    Merck Index 14,9644
    Health Hazard 1 Warning
    Odor Phenol-like
    Flash Point 99°C (210°F)
    RTECS Number SN1575000
    Quantity 5000g
    CAS 88-06-2
    UN Number UN2020
    Chemical Name or Material 2,4,6-Trichlorophenol
    Assay 98%

    Description

    Applications
    2,4,6-Trichlorophenol, antiseptic, defoliant, and glue preservative, chemical intermediate, anti-mildew treatment, solvent for polyester fiber.

    Solubility
    Soluble in water(0.8g/L).

    Notes
    Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

    GHS H Statement
    H351-H302-H315-H319
    Suspected of causing cancer.
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P280-P281-P305+P351+P338-P362-P405-P501a
    Wear protective gloves/protective clothing/eye protection/face protection.
    Use personal protective equipment as required.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Take off contaminated clothing and wash before reuse.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit