Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,4,6-Trichloropyrimidine-5-carboxaldehyde, 97%

Click to view available options
Quantity:
1g
5g
50270-27-4
C5HCl3N2O
211.426
MFCD02257700
KVJIRFGNHAAUNQ-UHFFFAOYSA-N
2,4,6-trichloro-5-pyrimidinecarboxaldehyde,2,4,6-trichloro-pyrimidine-5-carbaldehyde,2,4,6-trichloro-5-formylpyrimidine,5-formyl-2,4,6-trichloropyrimidine,2,4,6-trichloro-pyrimidine-5-carbal,2,4,6-trichloropyrimidine-5-carboxaldehyde,5-pyrimidinecarboxaldehyde, 2,4,6-trichloro,5-pyrimidinecarboxaldehyde,2,4,6-trichloro,pubchem13189,2,4,6-tri-chloropyrimidine-5-carbaldehyde
10932746
2,4,6-trichloropyrimidine-5-carbaldehyde
C(=O)C1=C(N=C(N=C1Cl)Cl)Cl
50270-27-4
C5HCl3N2O
211.426
MFCD02257700
KVJIRFGNHAAUNQ-UHFFFAOYSA-N
2,4,6-trichloro-5-pyrimidinecarboxaldehyde,2,4,6-trichloro-pyrimidine-5-carbaldehyde,2,4,6-trichloro-5-formylpyrimidine,5-formyl-2,4,6-trichloropyrimidine,2,4,6-trichloro-pyrimidine-5-carbal,2,4,6-trichloropyrimidine-5-carboxaldehyde,5-pyrimidinecarboxaldehyde, 2,4,6-trichloro,5-pyrimidinecarboxaldehyde,2,4,6-trichloro,pubchem13189,2,4,6-tri-chloropyrimidine-5-carbaldehyde
10932746
2,4,6-trichloropyrimidine-5-carbaldehyde
C(=O)C1=C(N=C(N=C1Cl)Cl)Cl
Applications
It is used as pharmaceutical intermediate. It is a versatile intermediate in the synthesis of reactive dyes.

Solubility
Slightly soluble in water.

Notes
Keep container tightly closed. Store in cool, well ventilated area. Store cold 2-4°C.
Quantity 5g
Color White to Brown
Melting Point 131°C to 132°C
UN Number UN2811
Beilstein 744505
Formula Weight 211.43
Physical Form Solid
Percent Purity 97%
Chemical Name or Material 2,4,6-Trichloropyrimidine-5-carboxaldehyde

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

storageNote1 : -30° to -10°C