Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4,6-Trichloropyrimidine 99%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
3764-01-0
C4HCl3N2
183.42
MFCD00006063
DPVIABCMTHHTGB-UHFFFAOYSA-N
pyrimidine, 2,4,6-trichloro,2,4,6-trichloro pyrimidine,2,4,6-trichloro-pyrimidine,ccris 7597,2,4,6-trichlorpyrimidin,pubchem9310,acmc-1agpv,2,6-trichloropyrimidine,2.4.6-trichloropyrimidin,dsstox_cid_29143
77378
2,4,6-trichloropyrimidine
C1=C(N=C(N=C1Cl)Cl)Cl
3764-01-0
C4HCl3N2
183.42
MFCD00006063
DPVIABCMTHHTGB-UHFFFAOYSA-N
pyrimidine, 2,4,6-trichloro,2,4,6-trichloro pyrimidine,2,4,6-trichloro-pyrimidine,ccris 7597,2,4,6-trichlorpyrimidin,pubchem9310,acmc-1agpv,2,6-trichloropyrimidine,2.4.6-trichloropyrimidin,dsstox_cid_29143
77378
2,4,6-trichloropyrimidine
C1=C(N=C(N=C1Cl)Cl)Cl
Boiling Point 210°C to 215°C
Melting Point 23°C to 25°C
Flash Point >112°C
Refractive Index 1.5690 to 1.571
Infrared Spectrum Authentic
Quantity 100g
Color Colorless to Yellow
Packaging Glass bottle
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Beilstein 23, 90
Formula Weight 183.42
Physical Form Liquid After Melting
Percent Purity ≥98.5%
Chemical Name or Material 2, 4, 6-Trichloropyrimidine, 99%
Show More Show Less

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 223-183-0

TSCA : TSCA