Learn More
2,4,6-Trifluoroaniline, 98%, Thermo Scientific Chemicals
Description
2,4,6-Trifluoroaniline was used in the synthesis of 3-nitro-2,4,6 -trifluoroacetanilide; eries of N?-phenyl-N-(1- phenyl cyclopentyl)-methyl ureas and 4-substituted 2,6-difluoro N-aryl pyridinones.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
2,4,6-Trifluoroaniline was used in the synthesis of 3-nitro-2,4,6 -trifluoroacetanilide; eries of N′-phenyl-N-(1- phenyl cyclopentyl)-methyl ureas and 4-substituted 2,6-difluoro N-aryl pyridinones.
Solubility
Insoluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
Melting Point | 33°C to 36°C |
Boiling Point | 57°C (22 mmHg) |
Flash Point | 54°C (129°F) |
Odor | Recognizable |
Quantity | 5 g |
UN Number | UN2926 |
Beilstein | 2831022 |
Solubility Information | Insoluble in water. |
SMILES | C1=C(C=C(C(=C1F)N)F)F |
Molecular Weight (g/mol) | 147.1 |
Show More |
Safety and Handling
Hazard Category | H228-H302+H312-H315-H318 |
Hazard Statement | GHS H Statement H331-H228-H373-H302-H312-H315-H319 Toxic if inhaled. Flammable solid. May cause damage to organs through prolonged or repeated exposure. Harmful if swallowed. Harmful in contact with skin. Causes skin irritation. Causes serious eye irritation. |
Precautionary Statement | P210-P240-P241-P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P310-P312-P330-P332+P313-P362-P370+P378q-P501c |
DOTInformation | Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, TOXIC, ORGANIC, N.O.S. |
EINECSNumber | 206-660-8 |
TSCA | No |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.