Alfa Aesar™ 2,4,6-Trifluoroaniline, 98%

Manufacturer: Alfa Aesar™ A1096403

Quantity 1g

 View more versions of this product

Catalog No. AAA1096403

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 363-81-5
    Molecular Formula C6H4F3N
    Formula Weight 147.1
    MDL Number MFCD00007650
    UN Number UN2926
    Beilstein 2831022
    Chemical Name or Material 2,4,6-Trifluoroaniline

    Specifications

    Formula Weight 147.1
    Health Hazard 2 GHS H Statement
    H331-H228-H373-H302-H312-H315-H319
    Toxic if inhaled.
    Flammable solid.
    May cause damage to organs through prolonged or repeated exposure.
    Harmful if swallowed.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.
    Health Hazard 3 GHS P Statement
    P210-P260-P261-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00007650
    CAS 363-81-5
    UN Number UN2926
    Boiling Point 57°C (22mmHg)
    Quantity 1g
    Odor Recognizable
    Molecular Formula C6H4F3N
    Health Hazard 1 Danger
    Melting Point 33° to 36°C
    Beilstein 2831022
    DOT Information Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, TOXIC, ORGANIC, N.O.S.
    Flash Point 54°C (129°F)
    Chemical Name or Material 2,4,6-Trifluoroaniline
    Assay 98%

    Description

    Applications
    2,4,6-Trifluoroaniline was used in the synthesis of 3-nitro-2,4,6 -trifluoroacetanilide; eries of N ft.-phenyl-N-(1- phenyl cyclopentyl)-methyl ureas and 4-substituted 2,6-difluoro N-aryl pyridinones.

    Solubility
    Insoluble in water.

    Notes
    Stable under recommended storage conditions. Incompatible with oxidizing agents.

    GHS H Statement
    H331-H228-H373-H302-H312-H315-H319
    Toxic if inhaled.
    Flammable solid.
    May cause damage to organs through prolonged or repeated exposure.
    Harmful if swallowed.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P210-P260-P261-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit