Alfa Aesar™ 2,4,6-Trifluoroanisole, 97%

Manufacturer: Alfa Aesar™ H2685206

Quantity 5g

 View more versions of this product

Catalog No. AAH2685206

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 219998-30-8
    Molecular Formula C7H5F3O
    Formula Weight 162.11
    MDL Number MFCD01631502
    UN Number UN3271
    Synonym 2-Methoxy-1,3,5-trifluorobenzene
    Chemical Name or Material 2,4,6-Trifluoroanisole

    Specifications

    Formula Weight 162.11
    Flash Point 35°C (95°F)
    Quantity 5g
    Molecular Formula C7H5F3O
    MDL Number MFCD01631502
    CAS 219998-30-8
    UN Number UN3271
    Synonym 2-Methoxy-1,3,5-trifluorobenzene
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H226
    Flammable liquid and vapor.
    Health Hazard 3 GHS P Statement
    P210-P280-P240-P303+P361+P353-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ETHERS, N.O.S.
    Chemical Name or Material 2,4,6-Trifluoroanisole
    Assay 97%

    GHS H Statement
    H226
    Flammable liquid and vapor.

    GHS P Statement
    P210-P280-P240-P303+P361+P353-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit