Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4,6-Trifluorobenzonitrile, 99%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
96606-37-0
C7H2F3N
157.095
MFCD00042399
HTKFGTCCOJIUIK-UHFFFAOYSA-N
benzonitrile, 2,4,6-trifluoro,2,4,6-trifluorobenzenecarbonitrile,ncr bf df ff,pubchem1565,acmc-209s7e,ksc488g9f,2,4,6-trifluoro-benzonitrile,2,4,6-trifluorobenzonitrile,benzonitrile, 2,4,6-trifluoro-, radical ion 1-9ci
737177
2,4,6-trifluorobenzonitrile
C1=C(C=C(C(=C1F)C#N)F)F
96606-37-0
C7H2F3N
157.095
MFCD00042399
HTKFGTCCOJIUIK-UHFFFAOYSA-N
benzonitrile, 2,4,6-trifluoro,2,4,6-trifluorobenzenecarbonitrile,ncr bf df ff,pubchem1565,acmc-209s7e,ksc488g9f,2,4,6-trifluoro-benzonitrile,2,4,6-trifluorobenzonitrile,benzonitrile, 2,4,6-trifluoro-, radical ion 1-9ci
737177
2,4,6-trifluorobenzonitrile
C1=C(C=C(C(=C1F)C#N)F)F
Packaging Glass bottle
Quantity 1g
Color White
Melting Point 57.0°C to 61.0°C
Infrared Spectrum Authentic
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula F3C6H2CN
Formula Weight 157.09
Physical Form Solid
Percent Purity 99%
Chemical Name or Material 2, 4, 6-Trifluorobenzonitrile, 99%
Show More Show Less

GHS H Statement:
Harmful if inhaled.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

TSCA : TSCA