Alfa Aesar™ 2,4,6-Trifluorophenylhydrazine, 97%

Manufacturer: Alfa Aesar™ H26104MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH26104MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 80025-72-5
    Molecular Formula C6H5F3N2
    Formula Weight 162.11
    MDL Number MFCD03095329
    Chemical Name or Material 2,4,6-Trifluorophenylhydrazine

    Specifications

    Formula Weight 162.11
    Molecular Formula C6H5F3N2
    MDL Number MFCD03095329
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H302-H315-H319-H335
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Melting Point 97° to 101°C
    Quantity 250mg
    CAS 80025-72-5
    Health Hazard 3 GHS P Statement
    P280h-P305+P351+P338
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Chemical Name or Material 2,4,6-Trifluorophenylhydrazine
    Assay 97%

    Description

    Applications
    Propionic acid (2,4,6-trichlorophenyl)hydrazide was prepared by reaction of (2,4,6-trichlorophenyl)- hydrazine with propionyl chloride in pyridine. Visnaginone Ia reacted with phenyl hydrazine and 2,4,6- trichlorophenyl hydrazine to yield the corresponding hydrazones.

    Solubility
    Insoluble in water.

    Notes
    Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

    GHS H Statement
    H302-H315-H319-H335
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P280h-P305+P351+P338
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit