Alfa Aesar™ 2,4,6-Trihydroxybenzoic acid monohydrate, 94%

Manufacturer: Alfa Aesar™ B2276836

Quantity 500g

 View more versions of this product

Catalog No. AAB2276836

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 71989-93-0
    Molecular Formula C7H6O5·H2O
    Formula Weight 188.14 (170.12 Anhydrous)
    MDL Number MFCD00149095
    Beilstein 2212148
    Synonym Phloroglucinolcarboxylic acid monohydrate
    Chemical Name or Material 2,4,6-Trihydroxybenzoic acid monohydrate

    Specifications

    Formula Weight 188.14 (170.12 Anhydrous)
    TSCA TSCA
    MDL Number MFCD00149095
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Melting Point ≈210°C (dec.)
    Quantity 500g
    Molecular Formula C7H6O5·H2O
    CAS 71989-93-0
    Beilstein 2212148
    Synonym Phloroglucinolcarboxylic acid monohydrate
    Health Hazard 1 Warning
    RTECS Number DH8910000
    Chemical Name or Material 2,4,6-Trihydroxybenzoic acid monohydrate
    Assay 94%

    Description

    Applications
    2,4,6-Trihydroxybenzoic acid monohydrate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

    Solubility
    Soluble in water.

    Notes
    Store in cool, dry conditions in a sealed container. Incompatible with oxidizing agents.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit