Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,4,6-Trihydroxybenzoic acid monohydrate, 94%

Click to view available options
Quantity:
25g
100g
500g
71989-93-0
C7H8O6
188.135
MFCD00149095
HWZIRFCGHAROOI-UHFFFAOYSA-N
2,4,6-trihydroxybenzoic acid hydrate,2,4,6-trihydroxybenzoic acid monohydrate,benzoic acid, 2,4,6-trihydroxy-, monohydrate,phloroglucinolcarboxylic acid,pubchem20337,acmc-209olb,2,4,6-trihydroxybenzoicacidmonohydrate,2,4,6-trihydroxybenzoic acid, hydrate,2,4,6-trihydroxybenzoic acid monohydrate, technical t,2,4,6-trihydroxybenzoic acid monohydrate, technical grade, predominantly 1,3,5-benzenetriol
2723793
2,4,6-trihydroxybenzoic acid;hydrate
C1=C(C=C(C(=C1O)C(=O)O)O)O.O
71989-93-0
C7H8O6
188.135
MFCD00149095
HWZIRFCGHAROOI-UHFFFAOYSA-N
2,4,6-trihydroxybenzoic acid hydrate,2,4,6-trihydroxybenzoic acid monohydrate,benzoic acid, 2,4,6-trihydroxy-, monohydrate,phloroglucinolcarboxylic acid,pubchem20337,acmc-209olb,2,4,6-trihydroxybenzoicacidmonohydrate,2,4,6-trihydroxybenzoic acid, hydrate,2,4,6-trihydroxybenzoic acid monohydrate, technical t,2,4,6-trihydroxybenzoic acid monohydrate, technical grade, predominantly 1,3,5-benzenetriol
2723793
2,4,6-trihydroxybenzoic acid;hydrate
C1=C(C=C(C(=C1O)C(=O)O)O)O.O
Applications
2,4,6-Trihydroxybenzoic acid monohydrate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Soluble in water.

Notes
Store in cool, dry conditions in a sealed container. Incompatible with oxidizing agents.
Quantity 25g
Melting Point ~210°C (decomposition)
Beilstein 2212148
Formula Weight 188.14 (170.12 Anhydrous)
Percent Purity 94%
Chemical Name or Material 2,4,6-Trihydroxybenzoic acid monohydrate

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : DH8910000

TSCA : TSCA